Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177353.3 | NP_796327.1 | 1 - 2 | 405 | 75 | |
GenBank mRNA | AK041169.1 | - | 1 - 2 | 405 | 75 | |
AK043187.1 | - | 1 - 2 | 427 | 75 | ||
AK078577.1 | - | 1 - 2 | 450 | 75 | ||
BC058750.1 | - | 1 - 2 | 411 | 6390297 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |