Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177353.3 | NP_796327.1 | 4 - 5 | 750 | 96 | |
GenBank mRNA | AK041169.1 | - | 4 - 5 | 750 | 96 | |
AK078577.1 | - | 4 - 5 | 795 | 96 | ||
BC058750.1 | - | 4 - 5 | 756 | 6390297 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |