Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145628.4 | NP_663603.3 | 3 - 4 | 448 | 65 | |
GenBank mRNA | AK031767.1 | - | 3 - 4 | 427 | 65 | |
AK049053.1 | - | 4 - 5 | 469 | 65 | ||
AK166523.1 | - | 3 - 4 | 424 | 65 | ||
AK192805.1 | - | 1 - 2 | 81 | 65 | ||
AK220478.1 | - | 2 - 3 | 161 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Mus musculus Probe spans exons |