Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011159.2 | NP_035289.2 | 41 - 42 | 5588 | 79 | |
GenBank mRNA | AB007544.1 | - | 41 - 42 | 5565 | 79 | |
AB011543.1 | - | 41 - 42 | 5577 | 79 | ||
D87521.1 | - | 41 - 42 | 5565 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |