Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011159.2 | NP_035289.2 | 79 - 80 | 11411 | 138 | |
GenBank mRNA | AB007544.1 | - | 79 - 80 | 11388 | 138 | |
AB011543.1 | - | 79 - 80 | 11400 | 138 | ||
AK088981.1 | - | 4 - 5 | 538 | 138 | ||
AK137732.1 | - | 6 - 7 | 845 | 138 | ||
D83786.1 | - | 4 - 5 | 546 | 138 | ||
D87521.1 | - | 79 - 80 | 11388 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |