Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163263.1 | NP_001156735.1 | 7 - 8 | 1376 | 68 | |
NM_182999.2 | NP_892044.1 | 8 - 9 | 1223 | 68 | ||
GenBank mRNA | AK048862.1 | - | 7 - 8 | 1335 | 68 | |
AK051278.1 | - | 7 - 8 | 1335 | 68 | ||
AK076501.1 | - | 3 - 4 | 305 | 68 | ||
AK139883.1 | - | 5 - 6 | 751 | 68 | ||
AK163905.1 | - | 4 - 5 | 588 | 68 | ||
AK220492.1 | - | 7 - 8 | 1292 | 68 | ||
BC004593.1 | - | 3 - 4 | 450 | 68 | ||
BC052482.1 | - | 8 - 9 | 1162 | 6308280 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |