Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175256.5 | NP_780465.4 | 10 - 11 | 2897 | 90 | |
GenBank mRNA | BC072651.1 | - | 2 - 3 | 207 | 90 | |
BC076596.1 | - | 8 - 9 | 2176 | 30645937 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |