Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199308.2 | NP_955012.2 | 9 - 10 | 1126 | 89 | |
GenBank mRNA | AK140821.1 | - | 9 - 10 | 1135 | 89 | |
AK155568.1 | - | 10 - 11 | 1098 | 89 | ||
CB524773.1 | - | 5 - 6 | 560 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |