Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012765.4 | NP_001012783.3 | 11 - 12 | 2817 | 78 | |
GenBank mRNA | AK147590.1 | - | 2920 | 78 | ||
AK147649.1 | - | 3577 | 78 | |||
AK160942.1 | - | 11 - 12 | 2817 | 78 | ||
BC035550.1 | - | 955 | 78 | |||
BC090846.1 | - | 11 - 12 | 2552 | 30363147 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |