Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012765.4 | NP_001012783.3 | 2 - 3 | 1748 | 104 | |
GenBank mRNA | AK147590.1 | - | 1849 | 104 | ||
AK147649.1 | - | 2508 | 104 | |||
AK160942.1 | - | 2 - 3 | 1748 | 104 | ||
BC090846.1 | - | 2 - 3 | 1483 | 30363147 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |