Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028610.2 | NP_082886.2 | 4 - 5 | 442 | 61 | |
GenBank mRNA | AK033197.1 | - | 4 - 5 | 443 | 61 | |
BC064754.1 | - | 4 - 5 | 381 | 30024281 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |