Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011173.2 | NP_035303.1 | 5 - 6 | 557 | 75 | |
GenBank mRNA | AK145501.1 | - | 5 - 6 | 557 | 75 | |
AK163032.1 | - | 5 - 6 | 585 | 75 | ||
BC128316.1 | - | 5 - 6 | 519 | 40109789 | 75 | |
L27439.1 | - | 4 - 5 | 368 | 75 | ||
Z25469.1 | - | 5 - 6 | 523 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |