Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008225.2 | NP_032251.2 | 1 - 2 | 92 | 62 | |
GenBank mRNA | AK040049.1 | - | 1 - 2 | 80 | 62 | |
AK088723.1 | - | 1 - 2 | 74 | 62 | ||
BC007469.1 | - | 1 - 2 | 62 | 3593456 | 62 | |
X84797.1 | - | 1 - 2 | 60 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |