Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172265.2 | NP_758469.1 | 7 - 8 | 1239 | 73 | |
GenBank mRNA | AK145801.1 | - | 7 - 8 | 1241 | 73 | |
AK160985.1 | - | 7 - 8 | 1168 | 73 | ||
BC027268.1 | - | 7 - 8 | 1186 | 73 | ||
BC038620.1 | - | 7 - 8 | 1239 | 4211776 | 73 | |
BC085255.1 | - | 7 - 8 | 1150 | 5342410 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |