Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159419.1 | NP_001152891.1 | 17 - 18 | 2031 | 66 | |
NM_198034.3 | NP_932151.2 | 17 - 18 | 2031 | 66 | ||
GenBank mRNA | AK034344.1 | - | 2 - 3 | 181 | 66 | |
AK046836.1 | - | 17 - 18 | 2391 | 66 | ||
AK171920.1 | - | 17 - 18 | 2184 | 66 | ||
AY562213.1 | - | 6 - 7 | 575 | 66 | ||
AY562214.1 | - | 6 - 7 | 575 | 66 | ||
BC025888.1 | - | 7 - 8 | 681 | 5039343 | 66 | |
BC079584.1 | - | 17 - 18 | 2031 | 5704672 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |