Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008909.2 | NP_032935.2 | 12 - 13 | 1528 | 55 | |
GenBank mRNA | AF126834.1 | - | 12 - 13 | 1468 | 55 | |
BC057152.1 | - | 12 - 13 | 1457 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |