Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008909.2 | NP_032935.2 | 20 - 21 | 2646 | 60 | |
GenBank mRNA | AF126834.1 | - | 20 - 21 | 2586 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |