Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028301.4 | NP_082577.2 | 6 - 7 | 827 | 72 | |
GenBank mRNA | AK012494.1 | - | 6 - 7 | 776 | 72 | |
AK089037.1 | - | 6 - 7 | 802 | 72 | ||
AK129471.1 | - | 6 - 7 | 771 | 72 | ||
BC050929.1 | - | 6 - 7 | 763 | 6807194 | 72 | |
BC051143.1 | - | 6 - 7 | 794 | 6531161 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |