Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027444.3 | NP_081720.2 | 13 - 14 | 2497 | 90 | |
GenBank mRNA | AF454943.1 | - | 13 - 14 | 2490 | 90 | |
AF454944.1 | - | 11 - 12 | 2362 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |