Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174848.3 | NP_777273.2 | 13 - 14 | 2922 | 91 | |
GenBank mRNA | AK030282.1 | - | 4 - 5 | 366 | 91 | |
AK162506.1 | - | 1 - 2 | 757 | 91 | ||
BC043118.1 | - | 13 - 14 | 2922 | 6408859 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |