Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081190.1 | NP_001074659.1 | 5 - 6 | 602 | 88 | |
GenBank mRNA | BC151105.1 | - | 5 - 6 | 652 | 9088018 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |