Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178923.3 | NP_849254.2 | 2 - 3 | 508 | 132 | |
GenBank mRNA | AK048239.1 | - | 2 - 3 | 570 | 132 | |
AK173120.1 | - | 2 - 3 | 586 | 132 | ||
BC053096.1 | - | 2 - 3 | 403 | 6401143 | 132 | |
BC057592.1 | - | 2 - 3 | 508 | 6417362 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |