Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161775.1 | NP_001155247.1 | 30 - 31 | 4261 | 59 | |
NM_013607.2 | NP_038635.2 | 30 - 31 | 4261 | 59 | ||
GenBank mRNA | AK137226.1 | - | 31 - 32 | 4238 | 59 | |
AK173045.1 | - | 30 - 31 | 4225 | 59 | ||
D85923.1 | - | 30 - 31 | 4216 | 59 | ||
D85924.1 | - | 30 - 31 | 4216 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |