Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039521.1 | NP_001034610.1 | 1 - 2 | 142 | 104 | |
GenBank mRNA | AK021410.1 | - | 1 - 2 | 134 | 104 | |
AK080397.1 | - | 1 - 2 | 97 | 104 | ||
AK080680.1 | - | 123 | 104 | |||
AK165676.1 | - | 1 - 2 | 131 | 104 | ||
BC138774.1 | - | 1 - 2 | 107 | 8861796 | 104 | |
BC138776.1 | - | 1 - 2 | 107 | 8861798 | 104 | |
CJ175904.1 | - | 1 - 2 | 145 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |