Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039521.1 | NP_001034610.1 | 7 - 8 | 650 | 86 | |
GenBank mRNA | AK080397.1 | - | 7 - 8 | 605 | 86 | |
AK080680.1 | - | 631 | 86 | |||
AK165676.1 | - | 7 - 8 | 639 | 86 | ||
BC034110.1 | - | 3 - 4 | 165 | 86 | ||
BC138774.1 | - | 7 - 8 | 615 | 8861796 | 86 | |
BC138776.1 | - | 7 - 8 | 615 | 8861798 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |