Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004063.mm.1 |
NM_001160402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004063.mm.1 |
NM_001160402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004063.mm.1 |
TC1600000440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004062.mm.1 |
NM_001160402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004062.mm.1 |
NM_001160402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004062.mm.1 |
TC1600000440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004064.mm.1 |
NM_001160402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004064.mm.1 |
NM_001160402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004064.mm.1 |
TC1600000440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004066.mm.1 |
NM_001160402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004066.mm.1 |
NM_001160402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004066.mm.1 |
TC1600000440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004065.mm.1 |
NM_001160402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004065.mm.1 |
NM_001160402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004065.mm.1 |
TC1600000440.mm.1 |
MTA TC |