Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199309.2 | NP_955013.1 | 7 - 8 | 909 | 71 | |
GenBank mRNA | AK132780.1 | - | 7 - 8 | 926 | 71 | |
AK133398.1 | - | 7 - 8 | 910 | 71 | ||
BC049761.1 | - | 7 - 8 | 909 | 6773873 | 71 | |
BC141059.1 | - | 6 - 7 | 818 | 9055671 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |