Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016924.2 | NP_058620.2 | 3 - 4 | 448 | 63 | |
GenBank mRNA | AF177771.1 | - | 3 - 4 | 404 | 63 | |
AK002746.1 | - | 3 - 4 | 414 | 63 | ||
AK084966.1 | - | 3 - 4 | 423 | 63 | ||
AK206976.1 | - | 1 - 2 | 67 | 63 | ||
BC002143.1 | - | 3 - 4 | 429 | 3484719 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |