Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081255.1 | NP_001074724.1 | 16 - 17 | 1796 | 72 | |
GenBank mRNA | AK134026.1 | - | 6 - 7 | 428 | 72 | |
AK155550.1 | - | 16 - 17 | 1794 | 72 | ||
BC151058.1 | - | 16 - 17 | 1774 | 9087971 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |