Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039530.3 | NP_001034619.2 | 12 - 13 | 4059 | 62 | |
GenBank mRNA | AK089685.1 | - | 13 - 14 | 4150 | 62 | |
AK220263.1 | - | 7 - 8 | 1651 | 62 | ||
BC047386.1 | - | 7 - 8 | 1160 | 62 | ||
BC138874.1 | - | 12 - 13 | 4174 | 8861896 | 62 | |
BC145452.1 | - | 12 - 13 | 4174 | 62 | ||
DQ372930.1 | - | 12 - 13 | 4223 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |