Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173026.2 | NP_766614.2 | 5 - 6 | 2052 | 140 | |
GenBank mRNA | AK033794.1 | - | 2 - 3 | 733 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |