Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081401.2 | NP_001074870.1 | 8 - 9 | 1657 | 64 | |
NM_177872.2 | NP_808540.2 | 8 - 9 | 1657 | 64 | ||
GenBank mRNA | BC158091.1 | - | 8 - 9 | 1258 | 9088176 | 64 |
BC158105.1 | - | 8 - 9 | 1258 | 9088181 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |