Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081401.2 | NP_001074870.1 | 9 - 10 | 1795 | 77 | |
NM_177872.2 | NP_808540.2 | 9 - 10 | 1795 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Mus musculus Probe spans exons |