Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026940.4 | NP_081216.1 | 2 - 3 | 332 | 76 | |
GenBank mRNA | AK007503.1 | - | 2 - 3 | 321 | 76 | |
AK032396.1 | - | 362 | 76 | |||
AK140044.1 | - | 437 | 76 | |||
AK141482.1 | - | 1 | 407 | 281 | ||
BC030425.1 | - | 2 - 3 | 334 | 76 | ||
BC115584.1 | - | 2 - 3 | 320 | 40102346 | 76 | |
BC115585.1 | - | 2 - 3 | 320 | 40102347 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |