Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176833.4 | NP_789803.1 | 1 - 2 | 154 | 96 | |
GenBank mRNA | AK016942.1 | - | 1 - 2 | 155 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |