Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138583.2 | NP_613049.2 | 4 - 5 | 337 | 75 | |
GenBank mRNA | AK089239.1 | - | 4 - 5 | 319 | 75 | |
AK133588.1 | - | 4 - 5 | 320 | 75 | ||
AK168595.1 | - | 3 - 4 | 316 | 75 | ||
BC005445.1 | - | 4 - 5 | 398 | 3496749 | 75 | |
X74504.1 | - | 3 - 4 | 310 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |