Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600001969.mm.1 |
NM_001166164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001969.mm.1 |
NM_001166164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001969.mm.1 |
TC1600000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001973.mm.1 |
NM_001166164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001973.mm.1 |
NM_001166164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001973.mm.1 |
TC1600000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001974.mm.1 |
NM_001166164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001974.mm.1 |
NM_001166164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001974.mm.1 |
TC1600000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001966.mm.1 |
NM_001166164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001966.mm.1 |
NM_001166164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001966.mm.1 |
TC1600000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001972.mm.1 |
NM_001166164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001972.mm.1 |
NM_001166164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001972.mm.1 |
TC1600000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001971.mm.1 |
NM_001166164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001971.mm.1 |
NM_001166164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001971.mm.1 |
TC1600000222.mm.1 |
MTA TC |