Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028429.1 | 3 - 4 | 949 | 96 | ||
GenBank mRNA | AK015967.1 | - | 4 - 5 | 1186 | 96 | |
AK039179.1 | - | 2 - 3 | 825 | 96 | ||
AK080650.1 | - | 2 - 3 | 824 | 96 | ||
BC089377.1 | - | 3 - 4 | 498 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |