Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026557.4 | NP_080833.1 | 1 - 2 | 441 | 64 | |
GenBank mRNA | AF071222.1 | - | 1 - 2 | 113 | 64 | |
AF276959.1 | - | 1 - 2 | 111 | 64 | ||
AF305423.1 | - | 1 - 2 | 97 | 64 | ||
AK018364.1 | - | 1 - 2 | 108 | 64 | ||
AK018488.1 | - | 1 - 2 | 138 | 64 | ||
AK078397.1 | - | 1 - 2 | 225 | 64 | ||
BC023138.1 | - | 1 - 2 | 215 | 4008685 | 64 | |
BC057143.1 | - | 1 - 2 | 441 | 5249543 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |