Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uso1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019490.1 | NP_062363.1 | 21 - 22 | 2636 | 66 | |
GenBank mRNA | AF096868.1 | - | 20 - 21 | 2424 | 66 | |
AK168433.1 | - | 9 - 10 | 1167 | 66 | ||
BC005548.1 | - | 5 - 6 | 604 | 66 | ||
BC016069.1 | - | 21 - 22 | 2636 | 4480992 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |