Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011148.3 | NP_035278.1 | 8 - 9 | 783 | 89 | |
GenBank mRNA | AF023458.1 | - | 8 - 9 | 783 | 89 | |
BC027049.1 | - | 9 - 10 | 1090 | 5356685 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |