Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030711.4 | NP_109636.1 | 5 - 6 | 961 | 117 | |
GenBank mRNA | AB047552.1 | - | 4 - 5 | 949 | 117 | |
AF227511.1 | - | 4 - 5 | 937 | 117 | ||
AK029841.1 | - | 5 - 6 | 960 | 117 | ||
AK030329.1 | - | 5 - 6 | 962 | 117 | ||
AK036559.1 | - | 962 | 117 | |||
AK040350.1 | - | 2 - 3 | 179 | 117 | ||
AK081181.1 | - | 5 - 6 | 988 | 117 | ||
AK159406.1 | - | 5 - 6 | 962 | 117 | ||
AK163954.1 | - | 5 - 6 | 981 | 117 | ||
BC046610.1 | - | 5 - 6 | 967 | 6397585 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |