Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025513.3 | NP_079789.1 | 1 - 2 | 335 | 69 | |
GenBank mRNA | AK006281.1 | - | 1 - 2 | 231 | 69 | |
AK012689.1 | - | 1 - 2 | 336 | 69 | ||
BC023669.1 | - | 1 - 2 | 330 | 5012335 | 69 | |
BC054085.1 | - | 1 - 2 | 340 | 3371550 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |