Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033201.3 | NP_001028373.2 | 8 - 9 | 1235 | 78 | |
GenBank mRNA | AK133991.1 | - | 8 - 9 | 1191 | 78 | |
AK134334.1 | - | 8 - 9 | 1226 | 78 | ||
AK166458.1 | - | 8 - 9 | 1017 | 78 | ||
BC033473.1 | - | 7 - 8 | 851 | 78 | ||
BC034658.1 | - | 5 - 6 | 677 | 78 | ||
BC055777.1 | - | 5 - 6 | 661 | 78 | ||
BC138898.1 | - | 8 - 9 | 1106 | 8861920 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |