Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293791.1 | NP_001280720.1 | 3 - 4 | 395 | 96 | |
NM_001293792.1 | NP_001280721.1 | 3 - 4 | 395 | 96 | ||
NM_027590.4 | NP_081866.1 | 3 - 4 | 395 | 96 | ||
GenBank mRNA | AK014939.1 | - | 3 - 4 | 371 | 96 | |
AK030974.1 | - | 3 - 4 | 359 | 97 | ||
AK034498.1 | - | 3 - 4 | 311 | 96 | ||
AK040574.1 | - | 3 - 4 | 396 | 96 | ||
AK047093.1 | - | 3 - 4 | 390 | 96 | ||
AK147743.1 | - | 3 - 4 | 369 | 96 | ||
AK169819.1 | - | 3 - 4 | 370 | 96 | ||
BC031923.1 | - | 3 - 4 | 358 | 4486323 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |