Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028182.1 | NP_082458.1 | 4 - 5 | 1051 | 73 | |
GenBank mRNA | AK008803.1 | - | 4 - 5 | 837 | 73 | |
AK157396.1 | - | 5 - 6 | 849 | 73 | ||
AX776203.1 | - | 3 - 4 | 508 | 73 | ||
BC023784.1 | - | 4 - 5 | 444 | 73 | ||
BC116682.1 | - | 4 - 5 | 759 | 40106612 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |