Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028182.1 | NP_082458.1 | 5 - 6 | 1169 | 66 | |
GenBank mRNA | AK008803.1 | - | 5 - 6 | 955 | 66 | |
AX776203.1 | - | 4 - 5 | 626 | 66 | ||
BC023784.1 | - | 5 - 6 | 562 | 66 | ||
BC116682.1 | - | 5 - 6 | 877 | 40106612 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |