Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008509.2 | NP_032535.2 | 3 - 4 | 629 | 78 | |
GenBank mRNA | AK002645.1 | - | 3 - 4 | 555 | 78 | |
AK017272.1 | - | 627 | 78 | |||
AK045064.1 | - | 3 - 4 | 518 | 78 | ||
AK079446.1 | - | 3 - 4 | 518 | 78 | ||
AK086023.1 | - | 3 - 4 | 450 | 78 | ||
AK150328.1 | - | 3 - 4 | 630 | 78 | ||
AK150355.1 | - | 3 - 4 | 630 | 78 | ||
AK150375.1 | - | 3 - 4 | 628 | 78 | ||
AK150457.1 | - | 3 - 4 | 525 | 78 | ||
AK150488.1 | - | 3 - 4 | 630 | 78 | ||
AK150505.1 | - | 3 - 4 | 630 | 78 | ||
AK150532.1 | - | 3 - 4 | 630 | 78 | ||
AK150577.1 | - | 3 - 4 | 630 | 78 | ||
AK150585.1 | - | 3 - 4 | 630 | 78 | ||
AK150593.1 | - | 3 - 4 | 630 | 78 | ||
AK150646.1 | - | 3 - 4 | 627 | 78 | ||
AK150672.1 | - | 3 - 4 | 548 | 78 | ||
AK150691.1 | - | 3 - 4 | 630 | 78 | ||
AK150984.1 | - | 3 - 4 | 630 | 78 | ||
AK150990.1 | - | 3 - 4 | 630 | 78 | ||
AK151006.1 | - | 3 - 4 | 630 | 78 | ||
AK151038.1 | - | 3 - 4 | 528 | 78 | ||
AK151049.1 | - | 3 - 4 | 630 | 78 | ||
AK151093.1 | - | 3 - 4 | 630 | 78 | ||
AK151099.1 | - | 3 - 4 | 630 | 78 | ||
AK151105.1 | - | 3 - 4 | 630 | 78 | ||
AK151112.1 | - | 3 - 4 | 631 | 78 | ||
AK151195.1 | - | 3 - 4 | 528 | 78 | ||
AK151216.1 | - | 3 - 4 | 631 | 78 | ||
AK151369.1 | - | 3 - 4 | 548 | 78 | ||
AK151434.1 | - | 3 - 4 | 548 | 78 | ||
AK151521.1 | - | 3 - 4 | 548 | 78 | ||
AK151540.1 | - | 3 - 4 | 518 | 78 | ||
AK151563.1 | - | 3 - 4 | 630 | 78 | ||
AK151630.1 | - | 3 - 4 | 528 | 78 | ||
AK151679.1 | - | 3 - 4 | 630 | 78 | ||
AK151698.1 | - | 3 - 4 | 630 | 78 | ||
AK151727.1 | - | 3 - 4 | 630 | 78 | ||
AK151772.1 | - | 3 - 4 | 630 | 78 | ||
AK151774.1 | - | 3 - 4 | 528 | 78 | ||
AK151801.1 | - | 3 - 4 | 518 | 78 | ||
AK151828.1 | - | 3 - 4 | 518 | 78 | ||
AK151866.1 | - | 3 - 4 | 630 | 78 | ||
AK151870.1 | - | 3 - 4 | 630 | 78 | ||
AK151872.1 | - | 3 - 4 | 630 | 78 | ||
AK151893.1 | - | 3 - 4 | 518 | 78 | ||
AK152014.1 | - | 3 - 4 | 630 | 78 | ||
AK152035.1 | - | 3 - 4 | 630 | 78 | ||
AK152049.1 | - | 3 - 4 | 630 | 78 | ||
AK152053.1 | - | 3 - 4 | 518 | 78 | ||
AK152079.1 | - | 3 - 4 | 630 | 78 | ||
AK152238.1 | - | 3 - 4 | 631 | 78 | ||
AK152251.1 | - | 3 - 4 | 628 | 78 | ||
AK152350.1 | - | 3 - 4 | 630 | 78 | ||
AK152390.1 | - | 3 - 4 | 469 | 78 | ||
AK152400.1 | - | 3 - 4 | 528 | 78 | ||
AK152481.1 | - | 3 - 4 | 631 | 78 | ||
AK152657.1 | - | 3 - 4 | 630 | 78 | ||
AK152706.1 | - | 3 - 4 | 631 | 78 | ||
AK152733.1 | - | 3 - 4 | 528 | 78 | ||
AK152775.1 | - | 3 - 4 | 630 | 78 | ||
AK152869.1 | - | 3 - 4 | 630 | 78 | ||
AK152956.1 | - | 3 - 4 | 630 | 78 | ||
AK153006.1 | - | 3 - 4 | 630 | 78 | ||
AK153148.1 | - | 3 - 4 | 630 | 78 | ||
AK153242.1 | - | 3 - 4 | 631 | 78 | ||
AK153425.1 | - | 3 - 4 | 630 | 78 | ||
AK153428.1 | - | 3 - 4 | 630 | 78 | ||
AK170486.1 | - | 3 - 4 | 630 | 78 | ||
BC003305.1 | - | 3 - 4 | 618 | 3584788 | 78 | |
BC158040.1 | - | 3 - 4 | 463 | 9088156 | 78 | |
CT010344.1 | - | 3 - 4 | 431 | 78 | ||
J03302.1 | - | 3 - 4 | 407 | 78 | ||
M65258.1 | - | 3 - 4 | 591 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |