Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172304.3 | NP_758508.2 | 5 - 6 | 1542 | 134 | |
GenBank mRNA | AK132870.1 | - | 5 - 6 | 1543 | 134 | |
AK141288.1 | - | 4 - 5 | 1321 | 134 | ||
BC006867.1 | - | 2 - 3 | 892 | 134 | ||
BC151176.1 | - | 4 - 5 | 1293 | 9088065 | 134 | |
BC151177.1 | - | 4 - 5 | 1293 | 9088066 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |