Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028902.1 | NP_083178.1 | 6 - 7 | 725 | 73 | |
GenBank mRNA | AK016553.1 | - | 6 - 7 | 710 | 73 | |
DV650538.1 | - | 6 - 7 | 725 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |